Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TG02 (Double bond E) 98+%

Supplier: AMBEED, INC
Warning

A440575-10MG A440575-1MG A440575-25MG A440575-5MG
77264-980EA 61 USD InStock
77264-980 77264-982 77264-984 77264-986
TG02 (Double bond E) 98+%
TG02 (Double bond E)
Precaución: for Research Use Only


Formula: C₂₃H₂₄N₄O
MDL Number: MFCD22124499
CAS Number: 1204918-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty