Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',6'-Dimethylacetanilide 96%

Supplier: AMBEED, INC
Warning

A441404-100G A441404-1G A441404-25G A441404-5G
77252-630EA 53 USD InStock
77252-630 77252-632 77252-634 77252-636
2',6'-Dimethylacetanilide 96%
2',6'-Dimethylacetanilide
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO
MDL Number: MFCD00008675
CAS Number: 2198-53-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty