Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diethylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A442099-100MG A442099-1G A442099-250MG
77240-964EA 22 USD InStock
77240-964 77240-966 77240-968
2,6-Diethylbenzoic acid 98%
2,6-Diethylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
MDL Number: MFCD09260802
CAS Number: 78114-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty