Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethyl]-1-phthalazinamine 98%

Supplier: AMBEED, INC
Warning

A442487-100MG A442487-1G A442487-250MG
77252-642EA 88 USD InStock
77252-642 77252-644 77252-646
(S)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethyl]-1-phthalazinamine 98%
(R,S)-N-PINAP
Precaución: for Research Use Only


Formula: C₃₈H₃₀N₃P
MDL Number: MFCD09264274
CAS Number: 828927-96-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty