Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2',3,3'-Tetrahydro-5,5'-bithieno[3,4-b][1,4]dioxine 95%

Supplier: AMBEED, INC
Warning

A444537-100MG A444537-1G A444537-250MG A444537-5G
77252-666EA 56 USD InStock
77252-666 77252-668 77252-670 77746-909
2,2',3,3'-Tetrahydro-5,5'-bithieno[3,4-b][1,4]dioxine 95%
2,2',3,3'-Tetrahydro-5,5'-bithieno[3,4-b][1,4]dioxine
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₄S₂
MDL Number: MFCD01325574
CAS Number: 195602-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty