Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromo-2,6-difluorophenyl)methanamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A446572-100MG A446572-1G A446572-250MG A446572-5G
77241-010EA 26 USD InStock
77241-010 77241-012 77241-014 77241-016
(4-Bromo-2,6-difluorophenyl)methanamine hydrochloride 97%
(4-Bromo-2,6-difluorophenyl)methanamine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₇BrClF₂N
MW: 258.49 g/mol
MDL Number: MFCD12546124
CAS Number: 1461655-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty