Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5(6)-Aminofluorescein (Mixture of isomers) 98% + mix

Supplier: AMBEED, INC
Warning

A446964-1G A446964-250MG A446964-5G
77265-056EA 112 USD InStock
77265-056 77265-058 77265-060
5(6)-Aminofluorescein (Mixture of isomers) 98% + mix
5(6)-Aminofluorescein (Mixture of isomers)
Precaución: for Research Use Only


Formula: C₂₀H₁₃NO₅
MW: 347.32 g/mol
MDL Number: MFCD00133331
CAS Number: 27599-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty