Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(4-methoxybenzoyl)benzoate 95%

Supplier: AMBEED, INC
Warning

A447792-100MG A447792-1G A447792-250MG A447792-5G
77291-654EA 11 USD InStock
77291-654 77291-656 77291-658 77291-660
Methyl 4-(4-methoxybenzoyl)benzoate 95%
Methyl 4-(4-methoxybenzoyl)benzoate
Precaución: for Research Use Only


Formula: C₁₆H₁₄O₄
MDL Number: MFCD07782837
CAS Number: 71616-84-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty