Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ferrocenium hexafluorophosphate 98%

Supplier: AMBEED, INC
Danger

A448548-100MG A448548-1G A448548-250MG
77265-090EA 23 USD InStock
77265-090 77265-092 77265-094
Ferrocenium hexafluorophosphate 98%
Ferrocenium hexafluorophosphate
Precaución: for Research Use Only


Formula: C₁₀H₁₀F₆FeP
MDL Number: MFCD00192173
CAS Number: 11077-24-0
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty