Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium cyclohexylmethyl trifluoroborate 98%

Supplier: AMBEED, INC
Warning

A451781-1G A451781-250MG
77241-038EA 31 USD InStock
77241-038 77241-040
Potassium cyclohexylmethyl trifluoroborate 98%
Potassium cyclohexylmethyl trifluoroborate
Precaución: for Research Use Only


Formula: C₇H₁₃BF₃·K
MDL Number: MFCD11052658
CAS Number: 1346647-18-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty