Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(6-Methoxynaphthalen-2-yl)propanoyl chloride 95%

Supplier: AMBEED, INC
Danger

A452255-100MG A452255-1G A452255-250MG
77252-798EA 117 USD InStock
77252-798 77252-800 77252-802
(S)-2-(6-Methoxynaphthalen-2-yl)propanoyl chloride 95%
(S)-2-(6-Methoxynaphthalen-2-yl)propanoyl chloride
Precaución: for Research Use Only


Formula: C₁₄H₁₃ClO₂
MDL Number: MFCD12031840
CAS Number: 51091-84-0
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty