Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl methylsulfonylacetate 97%

Supplier: AMBEED, INC
Warning

A456068-10G A456068-1G A456068-25G A456068-5G
77291-736EA 38 USD InStock
77291-736 77291-738 77291-740 77291-742
Ethyl methylsulfonylacetate 97%
Ethyl methylsulfonylacetate
Precaución: for Research Use Only


Formula: C₅H₁₀O₄S
MW: 166.20 g/mol
MDL Number: MFCD00041258
CAS Number: 4455-15-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty