Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: AMBEED, INC
Danger

A457002-1G A457002-250MG A457002-5G
77279-222EA 15 USD InStock
77279-222 77279-224 77279-226
3-Chloro-1H-pyrrolo[2,3-b]pyridine 97%
3-Chloro-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only


Formula: C₇H₅ClN₂
MDL Number: MFCD08272228
CAS Number: 80235-01-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty