Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

NSC-632839 98+%

Supplier: AMBEED, INC
Warning

A457204-10MG A457204-1MG A457204-25MG A457204-50MG A457204-5MG
77265-178EA 38 USD InStock
77265-178 77265-180 77265-182 77265-184 77265-186
NSC-632839 98+%
NSC-632839
Precaución: for Research Use Only


Formula: C₂₁H₂₂ClNO
MDL Number: MFCD09753281
CAS Number: 157654-67-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty