Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(hydroxymethyl)benzoate 95%

Supplier: AMBEED, INC
Warning

A461323-10G A461323-1G A461323-250MG
77279-278EA 392 USD InStock
77279-278 77279-280 77279-282
Methyl-3-(hydroxymethyl)benzoate 95%
Methyl 3-(hydroxymethyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
MDL Number: MFCD06202752
CAS Number: 67853-03-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty