Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Isopropoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A461585-10G A461585-1G A461585-250MG A461585-25G A461585-5G
77252-930EA 39 USD InStock
77252-930 77252-932 77252-934 77252-936 77252-938
3-Isopropoxyphenylboronic acid 98%
3-Isopropoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₃BO₃
MDL Number: MFCD03701512
CAS Number: 216485-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty