Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrothioanisole 98%

Supplier: AMBEED, INC
Warning

A461795-100G A461795-25G A461795-500G A461795-5G
77291-792EA 96 USD InStock
77291-792 77291-794 77291-796 77291-798
4-Nitrothioanisole 98%
4-Nitrothioanisole
Precaución: for Research Use Only


Formula: C₇H₇NO₂S
MW: 169.2
MDL Number: MFCD00010868
CAS Number: 701-57-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty