Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Ethylmorpholine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A461811-100MG A461811-1G A461811-250MG
77265-228EA 43 USD InStock
77265-228 77265-230 77265-232
(S)-2-Ethylmorpholine hydrochloride 97%
(S)-2-Ethylmorpholine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄ClNO
MDL Number: MFCD22631467
CAS Number: 1432794-09-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty