Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Proflavine hydrochloride (3,6-diaminoacridine hydrochloride) 98%

Supplier: AMBEED, INC
Warning

A465910-10G A465910-1G A465910-5G
77279-316EA 53 USD InStock
77279-316 77279-318 77279-320
Proflavine hydrochloride (3,6-diaminoacridine hydrochloride) 98%
Proflavine hydrochloride (3,6-diaminoacridine hydrochloride)
Precaución: for Research Use Only


Formula: C₁₃H₁₁N₃·HCl
MW: 245.71 g/mol
MDL Number: MFCD00066867
CAS Number: 952-23-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty