Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridin-2-ol 99+%

Supplier: AMBEED, INC
Danger

A467362-10G A467362-25G A467362-5G
77265-280EA 7 USD InStock
77265-280 77265-282 77265-284
Pyridin-2-ol 99+%
Pyridin-2-ol
Precaución: for Research Use Only


Formula: C₅H₅NO
MDL Number: MFCD00006268
CAS Number: 72762-00-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty