Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyanobenzoyl chloride 95%

Supplier: AMBEED, INC
Danger

A470238-10G A470238-1G A470238-250MG A470238-25G A470238-5G
77279-340EA 234 USD InStock
77279-340 77279-342 77279-344 77279-346 77279-348
3-Cyanobenzoyl chloride 95%
3-Cyanobenzoyl chloride
Precaución: for Research Use Only


Formula: C₈H₄ClNO
MW: 165.58
MDL Number: MFCD00011545
CAS Number: 1711-11-1
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty