Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-6,6'-dimethoxy-1,1'-biphenyl 98%

Supplier: AMBEED, INC
Warning

A470352-100MG A470352-1G A470352-250MG
77253-040EA 209 USD InStock
77253-040 77253-042 77253-044
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-6,6'-dimethoxy-1,1'-biphenyl 98%
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-6,6'-dimethoxy-1,1'-biphenyl
Precaución: for Research Use Only


Formula: C₄₂H₄₀O₂P₂
MDL Number: MFCD09753006
CAS Number: 133545-24-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty