Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrophenyl octyl ether 98%

Supplier: AMBEED, INC
Warning

A471221-100G A471221-25G A471221-5G
77279-358EA 352.01 USD InStock
77279-358 77279-360 77279-362
2-Nitrophenyl octyl ether 98%
2-Nitrophenyl octyl ether
Precaución: for Research Use Only


Formula: C₁₄H₂₁NO₃
MDL Number: MFCD00014693
CAS Number: 37682-29-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty