Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-((2S,3R)-3-((tert-Butoxycarbonyl)amino)-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid 97%

Supplier: AMBEED, INC
Warning

A472273-100MG A472273-1G A472273-250MG A472273-50MG
77265-350EA 156 USD InStock
77265-350 77265-352 77265-354 77748-977
(S)-2-((2S,3R)-3-((tert-Butoxycarbonyl)amino)-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid 97%
(S)-2-((2S,3R)-3-((tert-Butoxycarbonyl)amino)-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid
Precaución: for Research Use Only


Formula: C₂₁H₃₂N₂O₆
MDL Number: MFCD31631159
CAS Number: 87304-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty