Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oridonin 99%

Supplier: AMBEED, INC
Warning

A472649-100MG A472649-10MG A472649-1MG A472649-5G
77291-858EA 47 USD InStock
77291-858 77291-860 77291-862 77748-981
Oridonin 99%
Oridonin
Precaución: for Research Use Only


Formula: C₂₀H₂₈O₆
MDL Number: MFCD00221762
CAS Number: 28957-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty