Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Di-tert-butylphosphino)biphenyl 97%

Supplier: AMBEED, INC
Warning

A472747-100G A472747-10G A472747-1G A472747-250MG A472747-25G A472747-5G A472747-500G
77291-870EA 244.01 USD InStock
77291-870 77291-872 77291-874 77291-876 77291-878 77291-880 77748-983
2-(Di-tert-butylphosphino)biphenyl 97%
2-(Di-tert-butylphosphino)biphenyl
Precaución: for Research Use Only


Formula: C₂₀H₂₇P
MDL Number: MFCD01862440
CAS Number: 224311-51-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty