Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-nitrophenol 98%

Supplier: AMBEED, INC
Warning

A472994-100G A472994-10G A472994-1G
77291-882EA 91 USD InStock
77291-882 77291-884 77291-886
2-Chloro-5-nitrophenol 98%
2-Chloro-5-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
MDL Number: MFCD01571825
CAS Number: 619-10-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty