Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Diphenylpyridine 95%

Supplier: AMBEED, INC
Warning

A473460-100MG A473460-250MG
77279-382EA 10 USD InStock
77279-382 77279-384
3,5-Diphenylpyridine 95%
3,5-Diphenylpyridine
Precaución: for Research Use Only


Formula: C₁₇H₁₃N
MDL Number: MFCD04114211
CAS Number: 92-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty