Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Ethoxycarbonyl)-5-nitrophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A475019-1G A475019-25G A475019-5G
77253-076EA 20 USD InStock
77253-076 77253-078 77253-080
3-(Ethoxycarbonyl)-5-nitrophenylboronic acid 98%
3-(Ethoxycarbonyl)-5-nitrophenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₀BNO₆
MW: 238.9898 g/mol
MDL Number: MFCD03788422
CAS Number: 850568-37-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty