Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Benzyl-3-(hydroxymethyl)morpholine 97%

Supplier: AMBEED, INC
Warning

A475393-1G A475393-250MG A475393-5G
77291-910EA 29 USD InStock
77291-910 77291-912 77291-914
N-Benzyl-3-(hydroxymethyl)morpholine 97%
N-Benzyl-3-(hydroxymethyl)morpholine
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO₂
MDL Number: MFCD04038651
CAS Number: 110167-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty