Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(1H-indol-4-yl)piperazine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A475502-1G A475502-250MG A475502-5G
77291-916EA 263 USD InStock
77291-916 77291-918 77291-920
tert-Butyl 4-(1H-indol-4-yl)piperazine-1-carboxylate 97%
tert-Butyl 4-(1H-indol-4-yl)piperazine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₇H₂₃N₃O₂
MDL Number: MFCD04973301
CAS Number: 252978-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty