Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(3-Chlorophenyl)-1H-1,2,3-triazole 97%

Supplier: AMBEED, INC
Warning

A475572-100MG A475572-1G A475572-250MG
77265-424EA 127 USD InStock
77265-424 77265-426 77265-428
5-(3-Chlorophenyl)-1H-1,2,3-triazole 97%
5-(3-Chlorophenyl)-1H-1,2,3-triazole
Precaución: for Research Use Only


Formula: C₈H₆ClN₃
MDL Number: MFCD00220464
CAS Number: 35225-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty