Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-oxocyclopentaneacetate 97%

Supplier: AMBEED, INC
Warning

A476440-10G A476440-1G A476440-5G A476440-100G
77279-432EA 109 USD InStock
77279-432 77279-434 77279-436 77748-987
Ethyl-2-oxocyclopentaneacetate 97%
Ethyl-2-oxocyclopentaneacetate
Precaución: for Research Use Only


Formula: C₉H₁₄O₃
MDL Number: MFCD00044715
CAS Number: 20826-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty