Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Isopropyl-2-(3-oxo-3,4-dihydroquinoxalin-2-yl)phenyl)thiophene-2-carboxamide 98%

Supplier: AMBEED, INC
Warning

A476468-10MG A476468-1MG A476468-25MG A476468-50MG A476468-5MG
77241-302EA 54 USD InStock
77241-302 77241-304 77241-306 77241-308 77241-310
N-(4-Isopropyl-2-(3-oxo-3,4-dihydroquinoxalin-2-yl)phenyl)thiophene-2-carboxamide 98%
N-(4-Isopropyl-2-(3-oxo-3,4-dihydroquinoxalin-2-yl)phenyl)thiophene-2-carboxamide
Precaución: for Research Use Only


Formula: C₂₂H₁₉N₃O₂S
MDL Number: MFCD25976601
CAS Number: 1404437-62-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty