Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Chloromethyl)benzoic acid 98%

Supplier: AMBEED, INC
Danger

A476913-100G A476913-25G A476913-5G
77241-312EA 40 USD InStock
77241-312 77241-314 77241-316
4-(Chloromethyl)benzoic acid 98%
4-(Chloromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
MDL Number: MFCD00002568
CAS Number: 1642-81-5
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty