Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4-Benzyl-2-(quinolin-2-yl)-4,5-dihydrooxazole ≥ 98% (97.5%ee)

Supplier: AMBEED, INC
Warning

A477515-100MG A477515-1G A477515-50MG A477515-250MG
77265-438EA 46 USD InStock
77265-438 77265-440 77265-442 77748-990
(S)-4-Benzyl-2-(quinolin-2-yl)-4,5-dihydrooxazole ≥ 98% (97.5%ee)
(S)-4-Benzyl-2-(quinolin-2-yl)-4,5-dihydrooxazole
Precaución: for Research Use Only


Formula: C₁₉H₁₆N₂O
MDL Number: MFCD31803338
CAS Number: 1252576-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty