Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-(4-carboxybenzyl)piperazine 95%

Supplier: AMBEED, INC
Warning

A477782-10G A477782-250MG A477782-25G A477782-1G
77291-934EA 282.99 USD InStock
77291-934 77291-936 77291-938 77748-991
1-Boc-4-(4-carboxybenzyl)piperazine 95%
1-Boc-4-(4-carboxybenzyl)piperazine
Precaución: for Research Use Only


Formula: C₁₇H₂₄N₂O₄
MDL Number: MFCD04974012
CAS Number: 479353-63-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty