Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-(trifluoromethyl)benzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A478038-10G A478038-1G A478038-25G A478038-5G
77241-320EA 49 USD InStock
77241-320 77241-322 77241-324 77241-326
2-Fluoro-6-(trifluoromethyl)benzyl alcohol 97%
2-Fluoro-6-(trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₆F₄O
MDL Number: MFCD00061168
CAS Number: 152211-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty