Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1-(4-fluorophenyl)-2-phenylethanone 95%

Supplier: AMBEED, INC
Warning

A479470-1G A479470-250MG A479470-25G A479470-5G
77265-466EA 20 USD InStock
77265-466 77265-468 77265-470 77265-472
2-Bromo-1-(4-fluorophenyl)-2-phenylethanone 95%
2-Bromo-1-(4-fluorophenyl)-2-phenylethanone
Precaución: for Research Use Only


Formula: C₁₄H₁₀BrFO
MDL Number: MFCD00699290
CAS Number: 88675-31-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty