Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AS-604850 98+%

Supplier: AMBEED, INC
Warning

A480501-10MG A480501-1MG A480501-25MG A480501-50MG A480501-5MG
77265-480EA 19 USD InStock
77265-480 77265-482 77265-484 77265-486 77265-488
AS-604850 98+%
AS-604850
Precaución: for Research Use Only


Formula: C₁₁H₅F₂NO₄S
MW: 285.2 g/mol
MDL Number: MFCD19705528
CAS Number: 648449-76-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty