Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitroisothiazole-3-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A481136-100MG A481136-250MG
77279-482EA 58 USD InStock
77279-482 77279-484
5-Nitroisothiazole-3-carboxylic acid 97%
5-Nitroisothiazole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₄H₂N₂O₄S
MDL Number: MFCD11042296
CAS Number: 36778-15-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty