Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-1-boc-piperidine-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A481463-100MG A481463-1G A481463-250MG
77279-498EA 14 USD InStock
77279-498 77279-500 77279-502
3-Amino-1-boc-piperidine-3-carboxylic acid 98%
3-Amino-1-Boc-piperidine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₂₀N₂O₄
MW: 244.2900 g/mol
MDL Number: MFCD03790945
CAS Number: 368866-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty