Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Aminoethyl)morpholine 98%

Supplier: AMBEED, INC
Danger

A483919-100G A483919-10G A483919-25G A483919-500G A483919-5G
77291-952EA 59 USD InStock
77291-952 77291-954 77291-956 77291-958 77291-960
N-(2-Aminoethyl)morpholine 98%
N-(2-Aminoethyl)morpholine
Precaución: for Research Use Only


Formula: C₆H₁₄N₂O
MDL Number: MFCD00006182
CAS Number: 2038-03-1
UN: 3259
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty