Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4S,5S)-Diethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A483956-100MG A483956-1G A483956-250MG A483956-25G A483956-5G
77265-512EA 5 USD InStock
77265-512 77265-514 77265-516 77265-518 77265-520
(4S,5S)-Diethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate 97%
(4S,5S)-Diethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₈O₆
MW: 246.25 g/mol
MDL Number: MFCD18785757
CAS Number: 73346-73-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty