Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl (4-iodophenyl)acetate 95%

Supplier: AMBEED, INC
Warning

A484464-10G A484464-1G A484464-250MG A484464-25G A484464-5G
77291-976EA 55 USD InStock
77291-976 77291-978 77291-980 77291-982 77291-984
Ethyl (4-iodophenyl)acetate 95%
Ethyl (4-iodophenyl)acetate
Precaución: for Research Use Only


Formula: C₁₀H₁₁IO₂
MDL Number: MFCD07779004
CAS Number: 15250-46-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty