Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfamerazine 98%

Supplier: AMBEED, INC
Warning

A485232-100MG A485232-1G A485232-25G A485232-5G
77279-532EA 5 USD InStock
77279-532 77279-534 77279-536 77279-538
Sulfamerazine 98%
Sulfamerazine
Precaución: for Research Use Only


Formula: C₁₁H₁₂N₄O₂S
MW: 264.30 g/mol
MDL Number: MFCD00023212
CAS Number: 127-79-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty