Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-tert-Butyl-4-methoxyphenol 98%

Supplier: AMBEED, INC
Warning

A485686-100G A485686-25G A485686-500G A485686-5G
77291-986EA 58 USD InStock
77291-986 77291-988 77291-990 77291-992
2-tert-Butyl-4-methoxyphenol 98%
2-tert-Butyl-4-methoxyphenol
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
MDL Number: MFCD00040484
CAS Number: 121-00-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty