Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-thiazolecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A486131-100G A486131-25G
77292-014EA 173 USD InStock
77292-014 77292-016 77292-018
2-Amino-4-thiazolecarboxylic acid 98%
2-Amino-4-thiazolecarboxylic acid
Precaución: for Research Use Only


Formula: C₄H₄N₂O₂S
MW: 144.15 g/mol
MDL Number: MFCD00859429
CAS Number: 40283-41-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty