Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichloroanisole 98%

Supplier: AMBEED, INC
Warning

A486468-10G A486468-1G A486468-25G A486468-5G
77279-548EA 21 USD InStock
77279-548 77279-550 77279-552 77279-554
3,4-Dichloroanisole 98%
3,4-Dichloroanisole
Precaución: for Research Use Only


Formula: C₇H₆Cl₂O
MW: 177.03
Melting Pt: -7
Density: 1.33
MDL Number: MFCD00070785
CAS Number: 36404-30-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty