Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy1,3-dioxolane 98% (by GC)

Supplier: AMBEED, INC
Danger

A486582-25G A486582-5G
77265-556EA 77 USD InStock
77265-556 77265-558
2-Methoxy1,3-dioxolane 98% (by GC)
2-Methoxy1,3-dioxolane
Precaución: for Research Use Only


Formula: C₄H₈O₃
MDL Number: MFCD00003208
CAS Number: 19693-75-5
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty